Learn More
Hach Company Dissolved Oxygen Reagent Set, Buret, 300 mL sample
Supplier: Hach Company 2351400
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For determination of Dissolved Oxygen by the Winkler Method (Azide Modification) and buret titration. Range: up to 10 mg/L as DO. Sample Size: 300-mL. Contains reagents for 100 tests.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)