Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ UniStore™ Reagent Bottle, Narrow Mouth, HDPE, 90 mL, Pack of 6
Click to view available options
Capacity: 90 mL
Description
This narrow mouth bottle has a capacity of 90 mL and a lined polypropylene lid.
- Constructed of high density polyethylene, the bottle is translucent, pliable, and has excellent strength
- Leakproof bottle designed to meet TSA 3-1-1 guidelines for domestic and international travel
- Lined polypropylene lid
- Translucent
- Leakproof
- Certificates of Analysis available upon request
Specifications
Closure Type | Screw Cap |
Closure Material | Polypropylene |
Capacity | 90 mL |
Capacity (Metric) | 90 mL |
Capacity (English) | 3 oz. |
Color | Clear |
Material | HDPE |
Quantity | 6/Pk. |
Sterility | Non-sterile |
Closure Included | Yes |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction