Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reagent Alcohol, Honeywell™
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%
$662.00 - $5606.00
Specifications
CAS | 64-17-5, 67-56-1, 67-63-0 |
---|---|
CAS Max % | 90.00%, 5.00%, 5.00% |
Boiling Point | 78.32°C.The physical data is that of the main component. |
Flash Point | 15°C.The physical data is that of the main component. |
Packaging | Metal Drum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-048-572
|
Honeywell Chemicals
SR090ZA4 |
4 L |
Case of 4 for $662.00
|
|
|||||
60-090-446
|
Honeywell Chemicals
SR090ZA204L |
204 L |
Each for $5,606.00
|
|
|||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
64-17-5, 67-56-1, 67-63-0 | |
78.32°C.The physical data is that of the main component. | |
Metal Drum | |
ACS general use solvents |
90.00%, 5.00%, 5.00% | |
15°C.The physical data is that of the main component. | |
Liquid, clear |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title