Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reagent Alcohol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, anhydrous, SDA 3A, 90 ±1%, Honeywell
Click to view available options
Quantity:
4 x 4 L
Description
- ACS certified
- Lot-to-lot consistency
- HPLC grade — controlled for
- UV absorbance, GC assay, water content and non-volatile residue
Specifications
CAS Max % | 5.00, 90.00 |
Grade | HPLC |
Density | 0.7847 g/cm3 |
Boiling Point | 78.32°C |
Quantity | 4 x 4 L |
Flash Point | 15°C.The physical data is that of the main component. |
Packaging | Poly-coated Bottle |
Physical Form | Liquid |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | H225+H319+H370 |
Precautionary Statement | P280+P270+P264+P260+P243+P242+P241+P240+P233+P210 |
Recommended Storage | Room Temperature |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction