Learn More
Fujifilm Healthcare Americas Corp L-Type Ldl-C Reagent 2
Supplier: Fujifilm Healthcare Americas Corp 99900504
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The L-Type LDL-C Reagent 2 is for use in the L-Type LDL-C assay. The L-Type LDL-C is an in vitro assay utilizing a selective protection method for the determination of low-density lipoprotein cholesterol in serum or plasma. Package size: 2 x 20 mL bottles. This product is For In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.