Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Ready Access Lockable Lockout Station With (4) Key Hooks, Yellow
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Ready Access Lockable Lockout Station With (4) Key Hooks, Yellow is made of thermformed polypropylene for rugged durability. A semi-transparent hinged cover with locking mechanism protects contents from harsh environments and prevents unauthorized access.
Supplier: Brady™ 105942
Safety Services Available
Overview
LOTO Assessments
Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Cabinet | |
Polypropylene | |
LOCKOUT STATION | |
6.75 in. | |
17 in. |
Yellow | |
6.75 x 17 x 17 in. | |
17 in. | |
1 Ea. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction