Learn More
Ace Glass, Inc. 5000mL, one-piece jacketed pressure reactor with #25 center, (4)#15, (1)#7 side necks, 28/15 ball joint inlet/outlet, complete with addition funnel, condenser, pressure manifold and stirring assembly
Supplier: Ace Glass, Inc. 6438255
5000mL one-piece jacketed pressure reactor with a #25 center neck, (4) #15 and (1) #7 side necks, and a 28/15 o-ring ball joint on the inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.