Learn More
Ace Glass, Inc. 500mL, two-piece pressure reactor system, complete with glass body, 60mm head with (1) #7 and (3) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies
Supplier: Ace Glass, Inc. 6423200
500mL, two-piece pressure reactor system, complete with glass body, 60mm head with (1) #7 and (3) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies