Learn More
Ace Glass, Inc. 3000mL pressure vessel body, one-piece, #25 center, (4) #15, (1)#7 side necks, round bottom
Supplier: Ace Glass, Inc. 643631
3000mL round bottom pressure vessel body. Vessel has a #25 Ace-Thred center neck, 4 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. Rated 45 PSI at 100C.