Learn More
Ace Glass, Inc. 1000mL one-piece pressure vessel body, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet
Supplier: Ace Glass, Inc. 643713
1000mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet.