Learn More
Ace Glass, Inc. Reactor body, glass only, photochem, 500mL
Supplier: Ace Glass, Inc. 786318
500mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Vessel includes a flat bottom that will accept the use of a stir magnet.