Learn More
Pluriselect Re-Strainer (In-Line-Strainer) 100 um mesh-size, 20 pcs. Sterile
Supplier: Pluriselect 437510040
The Re-Strainer (In-Line-Strainer) has a 6 ml recovery reservoir which can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports. These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor. You can choose from 15 different mesh sizes (1 um to 500 um)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.