Learn More
Rats, Fisher Science Education™ Fisher-Free™
SCI_ED
Rattus norvegicus.
$18.75 - $26.75
Specifications
Preservative Fisher-Free™
Quantity Ea.
View More Specs
Products 5
Catalog Number Mfr. No. Injection Type Type Price Quantity & Availability  
Catalog Number Mfr. No. Injection Type Type Price Quantity & Availability  
S1178S
SCI_ED
View Documents
Ranaco Corporation
140.1178.1
Plain Rat
Pack for $20.75
Add to Cart
S1180S
SCI_ED
View Documents
Ranaco Corporation
140.1180.1
Double Rat
Pack for $18.75
Add to Cart
S1182S
SCI_ED
View Documents
Ranaco Corporation
140.1182.1
Double Rat, Pregnant
Pack for $26.75
Add to Cart
S1181S
SCI_ED
View Documents
Ranaco Corporation
140.1181.1
Triple Rat
Pack for $21.75
Add to Cart
S1179S
SCI_ED
View Documents
Ranaco Corporation
140.1179.1
Single Rat
Pack for $22.00
Add to Cart
Description
Description
Rats are similar in price to fetal pigs, but have adult musculature. If you desire a mammal specimen that is inexpensive, fully muscled, and has well-developed genitalia, the rat is the specimen of choice. Pregnant rats are also available.
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue, and hepatic portal veins yellow
Specifications
Specifications
Fisher-Free™
Ea.