Learn More
Innovative Research Inc Innovative Grade US Origin Rat Wistar Complement Serum 5ml
Supplier:  Innovative Research Inc IGRTWICSER5ML
Encompass_Preferred
Innovative Grade US Origin Rat Wistar Complement Serum from Innovative Research is manufactured using whole blood collections from type Wistar rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity - custom volumes are available upon request. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-5783