Learn More
Sklar Instruments Tissue Forceps, Sklar , 6 in., 1x2 Teeth, Tissue Forceps, Straight, Stainless Steel, Reusable. Sklar Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. EA.
Supplier: Sklar Instruments 191260
Tissue Forceps, Sklar, 6 in., 1x2 Teeth, Tissue Forceps, Straight, Stainless Steel, Reusable. Sklar Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. EA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.