Learn More
Innovative Research Inc Rat Sprague Dawley Red Blood Cells 50ml
Supplier:  Innovative Research Inc IRTSDRBC50ML
Encompass_Preferred
Rat Sprague Dawley Red Blood Cells from Innovative Research are manufactured using whole blood collections from type Sprague Dawley rats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4886