Learn More
BioVendor, LLC Rat Quality Control Set (TSH, Testosterone, Corticosterone, Prolactin) (2 x 1 ml)
Supplier: BioVendor, LLC RTC900R
TypeCompetitive ELISA, Immobilized antigenApplicationsSerum, PlasmaSample Requirements10 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range15–2250ng/mlLimit of Detection6.1 ng/ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.