Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgG1 Kappa Light Chain Isotype Control (KLH/G1-2-2), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP151817FR
Description
IgG1 Kappa Light Chain Monoclonal specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.Specifications
IgG1 Kappa Light Chain | |
Monoclonal | |
DyLight 680 | |
50 mM Sodium Borate. with 0.05% Sodium Azide | |
Keyhole limpet hemocyanin (KLH) | |
RUO | |
Immunoglobulin reacts with Keyhole limpet hemocyanin (KLH). | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence | |
KLH/G1-2-2 | |
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
Rat | |
500 Tests | |
Isotype Control (Primary) | |
Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction