Learn More
Novus Biologicals™ Rat Creatine Kinase MB ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275313
Description
The Rat Creatine Kinase MB ELISA Kit (Colorimetric) measures rat Creatine Kinase MB in serum, plasma and other biological fluids.Specifications
31.25 to 2000 pg/mL | |
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed. | |
Other Biological Fluids, Plasma, Plasma, Serum, Serum | |
ELISA | |
CKB | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Creatine Kinase MB |
18.75 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed. | |
1152 | |
Rat | |
metabolism | |
Creatine Kinase Isoenzyme |