Learn More
Rat anti-Mouse IgG2a, clone: LO-MG2a, Secondary Antibody, Invitrogen™
Rat Monoclonal Secondary Antibody
Supplier: Thermo Scientific RMG2A00
Description
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgG2a | |
Monoclonal | |
Unconjugated | |
Rat | |
Purified | |
Secondary | |
Store at 4° C | |
IgG1 |
Immunofluorescence | |
LO-MG2a | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
Mouse IgG2a | |
500 μL | |
Mouse | |
Purified |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.