Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rainbow Calibration Particle Set (6 peaks)

Used for long term performance tracking or routine calibration of flow cytometers

Supplier:  Novus Biologicals™ NBP300505

Catalog No. NB112518


Add to Cart

Description

Description

Rainbow Calibration Particles are a mixture of fluorophore incorporated particles with 8 different fluorescent intensities that are used for calibrating flow cytometers
Specifications

Specifications

Fluorescence Imaging, Flow Cytometry
5 mL
0.02% Sodium Azide
Store at 4°C in the dark. Do not freeze.
Deionized water, 0.01% NP-40
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit