Learn More
Research Products International Corp Corrugated Radioactive Waste Box, 12 1/4 x 12 1/4 x 24 3/4 Inches, 25 per Case
SDP
Supplier:  Research Products International Corp BRS15
Encompass_Preferred
Heavy duty corrugated radioactive waste boxes have a radiation and caution warning symbols imprinted on the boxes. These can be used individually or with a Rad-4 Lock Box for a complete shielding waste system. Boxes ship flat to prevent damage and need to be folded into final shape before use.
Catalog No. 50-550-398