Learn More
Research Products International Corp Radioactive Waste Disposal Bags, 3 Mil Thick, 8 x 6 x 15 Inches, Yellow Tint, Pre-Printed with Caution Message, 100 per Package
SDP
Supplier:  Research Products International Corp 118615
Encompass_Preferred
Radioactive material waste bags are available in a variety of convenient sizes and styles to help sort and identify different types of waste. Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.
Catalog No. 50-550-148