Learn More
Research Products International Corp Radioactive Waste Disposal Bags, 3 Mil Thick, 11 x 11 x 30 Inches, Yellow Tint, Pre-Printed with Caution Message, 100 per Package
Supplier: Research Products International Corp 111130
Radioactive material waste bags are available in a variety of convenient sizes and styles to help sort and identify different types of waste. Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.