Learn More
Thermo Scientific™ RadEye™ GR Grapple Monitoring System
Minimize the threat of radioactive material in the scrap metal stream with a wireless radiation detection system designed for grapple installation.
$434.00
Specifications
Communication Details | ZigBee transmitter, 1 mW, >Range 100 m, 328 ft. |
---|---|
Data Management Details | Internal Data Logger; RadEye PC-software compliant |
Includes | System Components: RadEye R Receiver Unit (option to increase the number of Receiver Units) ; Detector Unit (Installed on grapple) ; Battery Module and Radio Transceiver (Installed on grapple) ; Optional Datalogger |
Interface Board | IR (standard) or Bluetooth (option) |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
19-175-648
|
Thermo Scientific™
425522005 |
Gooseneck adapter |
Each of 1 for $434.00
|
|
|||||
Description
In the metal recycling industry, discarded radioactive materials can seriously contaminate a shredder, baler, steel melt or bag house, resulting in significant clean-up costs and lost productivity. The Thermo Scientific™ RadEye™ GR Grapple Monitoring System is a wireless radiation detection system designed to be installed on a grapple to monitor scrap metal while it is still in the scrap pile under the grapple. Even when radiation is highly attenuated, the RadEye GR system responds to shielded radioactive sources to stop them from making their way into the steel making process.
The RadEye GR Grapple Monitoring System is a radiation detection device designed and proven through extensive testing for the extreme forces and harsh conditions experienced when installed in a grapple. The RadEye GR system provides an additional, significant security increase to present portal monitoring because it is closer to the radioactive source and maintains contact for a longer period of time.
Compact detector size - negligible load capacity reduction while providing high sensitivity.
Enhanced sensitive for low gamma energies.
Superior value: Small investment and low cost of ownership.
Multiple portable RadEye GR display units possible.
Extremely high battery lifetime.
Nearly maintenance-free.
Very straightforward installation process.
Simple and comprehensive data logging and reporting.
Radiation test handled by single person.
ViewPoint centralized real time monitoring option.
Optimized shape and thickness of the detector dome allows even low energetic radiation to cause a significant signal in the detector.
System Components:
RadEye R Receiver Unit (option to increase the number of Receiver Units).
Detector Unit (Installed on grapple).
Battery Module and Radio Transceiver (Installed on grapple).
Optional Datalogger.
Specifications
ZigBee transmitter, 1 mW, >Range 100 m, 328 ft. | |
Internal Data Logger; RadEye PC-software compliant | |
System Components: RadEye R Receiver Unit (option to increase the number of Receiver Units) ; Detector Unit (Installed on grapple) ; Battery Module and Radio Transceiver (Installed on grapple) ; Optional Datalogger | |
IR (standard) or Bluetooth (option) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.