Learn More
Se International Inc RAD 60 / Alarming Radiation Dosimeter
SDP
Supplier:  Se International Inc RADOS RAD60R
Encompass
The RADOS RAD-60 Personal Alarm Dosimeter is a precise and reliable instrument for ensuring the personal safety of the user. Ideally, the RAD-60 is used in everyday radiation monitoring, in stand-alone conditions. The RAD-60 can be switched into System Mode, for the purpose of tracking Personnel Dose records and generating compliance reports. The design includes state-of-the-art technology with built in memory for retrieving dose, even during power-down. It eliminates outside interference from shock and RF. The RAD-60 is easily programmed by the user, has a digital display, and operates with a single AAA alkaline battery. With the push of a button, you can turn the unit on/off, change the digital display to read dose or dose rate, select from several dose and dose rate alarm levels, turn the chirp function on/off, reset the integrated dose, and perform battery tests.The large digital display gives instant dose or dose rate readings and the audible alarm is loud (>85dba)
Catalog No. NC9969391