Learn More
Research Products International Corp Rad-Monitor, SD10
SDP
Supplier:  Research Products International Corp 954065
Encompass_Preferred
Model SD10 is supplied with a high sensitivity scintillation probe for detecting low energy gamma emitters such as Iodine-125. The detector utilizes a high quality photomultiplier tube coupled to a 1 Inch x 2 mm thick sodium iodide crystal with a thin 1 mil (7 mg/cm2) aluminum window. Also suitable for thyroid monitoring of 125I uptake.
Catalog No. 50-197-6931
Explore available promotions