Learn More
Research Products International Corp Radioactive Waste Disposal Bags, 4 Mil Thick, 33 x 40 Inches, Yellow Opaque, Unprinted, 25 per Package
SDP
Supplier:  Research Products International Corp 286947Y
Encompass_Preferred
Radioactive material waste bags help sort and identify different types of waste. Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.
Catalog No. 50-213-092