Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Racked Filtered Micropipette Tips

Protect samples and pipettes from contamination. Eisco™ Racked Filtered Micropipette Tips are designed to be used with variable or fixed single and multi channel micropipettes.
$38.50
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Autoclavable | Autoclavable |
For Use With (Application) | Electrophoresis, PCR |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
S39866
![]() |
Eisco™
CH0591N |
0.2 to 10 μL |
Each for $38.50
|
|
S39867
![]() |
Eisco™
CH0591O |
2 to 20 μL |
Each for $38.50
|
|
S39868
![]() |
Eisco™
CH0591P |
2 to 50 μL |
Each for $38.50
|
|
S39869
![]() |
Eisco™
CH0591Q |
2 to 100 μL |
Each for $38.50
|
|
S39870
![]() |
Eisco™
CH0591R |
2 to 200 μL |
Each for $38.50
|
|
S39871
![]() |
Eisco™
CH0591S |
50 to 300 μL |
Each for $38.50
|
|
S39872
![]() |
Eisco™
CH0591T |
200 to 1000 μL |
Each for $38.50
|
|
Description
- Sterile
- Graduated
- Contain hydrophobic polyethylene filter
- Free of lubricants, dyes and heavy metals
- 96 tips supplied in reusable racks
- Rack and tips are autoclavable
- Made of plastic
- Free of lubricants, dyes, and heavy metals
Specifications
Filtered | |
Clear | |
Electrophoresis, PCR | |
Rack of 96 | |
Micropipettes | |
PP (Polypropylene) |
Sterile | |
Autoclavable | |
Rack | |
Yes | |
Yes | |
Rectangle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title