Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Racked Filter Pipette Tips



These filter pipette tips are sterile, racked, and laboratory approved.
Supplier: United Scientific™ UNUFT01000UL
Description
- Protect against cross-contamination using these racked, sterile filter pipette tips
- These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipette
- These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE)
- DNase, RNase and Pyrogen-free
Specifications
1000 μL | |
Sterile | |
Autoclavable | |
Pipetting | |
Rack, 10 x 96 | |
Racked | |
Polypropylene |
Filtered | |
Clear | |
UNUFT01000UL | |
Universal | |
Standard Length | |
Pipettors |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title