Learn More
PATTERSON DENTAL COMPANY Racellet Hemostatic Cotton Pellets, 350/pk
Supplier: PATTERSON DENTAL COMPANY 5568985
Size: 3, 0.55 mg Epinephrine HCI, Hemostatic – use to control light bleeding and seepage along gingival margins. Pellets are activated by normal tissue moisture. For rapid activation, dip in water just prior to application.
This item is not returnable.
View return policy