Learn More
Rabbits, Bio-Fresh™
Oryctolagus cuniculus.
Supplier: Ranaco Corporation 140.1059.1
Description
Excellent for herbivore vs. carnivore comparative studies. Excellent herbivore specimen for study. We recommend dissecting at least one rabbit along with your normal cat study.
Strongest comparisons between the rabbit and cat anatomy exist within:
- Intestinal systems
- Eye position and visual acuity
- Dentition
- Forearm musculature and functionality
- Each rabbit comes with a tag and label identifying the sex
Order Info
INJECTION KEY:
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Skinned Rabbit | |
Bio-Fresh™ | |
Double | |
Ea. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.