Learn More
Rabbits, Bio-Fresh™
SCI_ED
Oryctolagus cuniculus.
 View more versions of this product
Catalog No. S1057S
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Description
Description
Excellent for herbivore vs. carnivore comparative studies. Excellent herbivore specimen for study. We recommend dissecting at least one rabbit along with your normal cat study.
Strongest comparisons between the rabbit and cat anatomy exist within:
  • Intestinal systems
  • Eye position and visual acuity
  • Dentition
  • Forearm musculature and functionality
  • Each rabbit comes with a tag and label identifying the sex
Order Info
  • INJECTION KEY:
  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
  • Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Specifications
Skinned Rabbit
Bio-Fresh™
Plain
Ea.
Product Suggestions
Product Suggestions