Learn More
Rockland Immunochemicals Rabbit Serum Sterile 100 mL
Supplier: Rockland Immunochemicals D109000100
Rabbit Serum is used as a supplement to cell culture media. Rabbit Serum is also suitable for use as a component of bioassays immunoassays or enzyme assays. Rabbit Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation. Rabbit Serum is ideal for investigators in Cancer Immunology and Cell Biology research.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.