Learn More
Bioworld Rabbit Serum, Non-Sterile, 100 mL
Supplier: Bioworld 306111571
Rabbit Serum : Collected from 8-12 week-old New Zealand or Californian rabbits. Aseptically collected and processed to minimize hemolytic activity.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.Raw, non-sterile Low hemoglobin levelsPhysiochemical constituents measuredto ensurelot to lot consistencyEndotoxin testing availableMycoplasma testing availableStorage conditions: -20° ± 5°C Country of origin: USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.