Learn More
Innovative Research Inc Rabbit Red Blood Cells 25ml
Supplier: Innovative Research Inc IRBRBC25ML
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Rabbit Red Blood Cells from Innovative Research are manufactured using whole blood collections from rabbits. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.