Learn More
Rockland Immunochemicals Rabbit Red Blood Cells 10% Washed Pooled Cells 50 mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Rockland Immunochemicals R4030050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Rabbit whole blood is washed to remove the platelet rich plasma buffy coat layer and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Rabbit red blood cells are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Rabbit red blood cells are perishable and are collected and processed upon receipt of your order.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.