Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit IgG Isotype Control, BD
Supplier: BD Biosciences 550875
Description
Host Species: Rabbit
Isotype: IgG
Applications:
ELISA Standard (Routinely Tested)
Immunohistochemistry, Flow cytometry (Not Recommended)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ELISA | |
Unconjugated | |
Trinitrophenol-KLH | |
0.5 mg | |
IgG |
0.5mg/mL | |
Rabbit | |
Affinity Purified | |
Undiluted at 4°C |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction