Learn More
Bioworld Rabbit Gamma Globulin Liquid >= 96%, 1 gram, 10mg/ml, 100ml, 1 g
Supplier: Bioworld 220701101
The gamma globulin preparations are supplied as a liquid. Purity levels as GG are analyzed by SDS-Page or Agarose electrophoresis.These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps,or as a protein stabilizer blocker in a number of immunoassays.These preparations are also useful as GG control solutions for biochemical analytical tests.Gamma globulin preparations are provided at pH 7.2 ± 0.2. Usage:bioWORLD\'s products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.