Learn More
Rockland Immunochemicals Rabbit Blood In Sodium Citrate 50 mL
SDP
Supplier:  Rockland Immunochemicals R2090050
Encompass_Preferred
Citrated rabbit blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Rabbit red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-194-3983