Learn More
NEW EAST BIOSCIENCES RAB11 PULL-DOWN ACTIVATION ASS
Supplier:  NEW EAST BIOSCIENCES 83201
Encompass_Preferred
Rab11 Pull-Down Activation Assay KitSize: 30 AssaysPositive Applications: Pull-DownMeasurment: Active Rab11-GTP level
Catalog No. NC2330731