Learn More
Enzo Life Sciences R-Spondin-1 (human), (recombinant) (20µg)
Supplier: Enzo Life Sciences ENZPRT1520020
Alternative name: Rspo-1, Roof plate-specific spondin-1. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 7.5, containing 150mM NaCl. Source: Produced in CHO cells. Long Term Storage: -20°C.