Learn More
R&D Systems™ Cultrex 3-D Spheroid Fluorometric Assay, 96-well
Offers an in vitro, standardized, 3D, high-content format for inducing multicellular tumor spheroid (MCTS) formation and quantitating cell viability in response to pharmacological treatment
Supplier: R&D Systems™ 3510096K
Includes: 10X Spheroid Formation ECM, 600uL; 3D Culture Qualified 96 Well Spheroid Formation Plate; Resazurin, 2.5mL
Description
Other in vitro tumor models lack either a physiological context and/or reproducible format for evaluating tumor cells in vitro. The most popular method for compound screening and pathway analysis involves culturing cancer cells on rigid, tissue culture tr
Uses a 3D Culture Qualified 96 Well Spheroid Formation Plate alongside a specialized Spheroid Formation ECM to drive aggregation and/or spheroid formation of cells
Upon completion of spheroid formation, the spheroid may be treated with pharmacological ag
Specifications
The unopened product requires storage at both -80°C and -20°C. Consult the product insert for specific storage temperatures. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Each |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.