Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Quik-Align™ No Water Picto Polyester Label
Easily apply outdoor-durable Quik-Align™ labels to identify sites, chemical containers or other points of emphasis where unusual reactivity with water could occur.
$10.50 - $28.75
Specifications
Material | Laminated Polyester |
---|---|
Quantity | 10 Labels/Pk. |
Adhesive Type | Permanent Acrylic |
For Use With (Application) | Facility Identification, Hazard Communication Identification, Safety Identification |
Temperature Range (Metric) | -40°C to +110°C |
Catalog Number | Mfr. No. | Height (English) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Width (English) | Price | Quantity & Availability | ||||
17-200-546
|
Brady™
175209 |
1 in. | 1.25 in. |
Pack of 10 for $10.50
|
|
||||
17-200-538
|
Brady™
175153 |
2 in. | 2.25 in. |
Pack of 10 for $10.75
|
|
||||
17-200-558
|
Brady™
175251 |
3 in. | 3.375 in. |
Pack of 10 for $13.50
|
|
||||
17-200-440
|
Brady™
175334 |
4 in. | 4.25 in. |
Pack of 10 for $18.50
|
|
||||
17-200-445
|
Brady™
175491 |
6.75 in. | 6.25 in. |
Pack of 10 for $28.75
|
|
||||
Description
- Card-mounted polyester film characters feature pressure-sensitive adhesive
- Flexible material conforms to most surfaces and holds up to harsh environments, abrasion, and repeated spills and cleanings
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (-40°F to +230°F)
Specifications
Laminated Polyester | |
Permanent Acrylic | |
-40°C to +110°C | |
W |
10 Labels/Pk. | |
Facility Identification, Hazard Communication Identification, Safety Identification | |
-40°F to +230°F | |
Black on White |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title