Learn More
Quidel Sofia™ Strep A FIA CLIA-Waived
SDP
For rapid qualitative detection of Group A Streptococcal antigens from throat swabs of symptomatic patients
Supplier:  Quidel 20281
 View more versions of this product
Catalog No. 23043085
Due to product restrictions, please to purchase or view availability for this product.
Includes: 25 Test-Kit
  • Individually Packaged Cassettes (25): Polyclonal rabbit anti-Group A Streptococcus antibodies
  • Reagent Tubes (25)
  • Reagent Solution Bottles (25): 4M Sodium Nitrite and 0.2M Acetic Acid inside glass ampoule
  • Fixed Volume Pipettes (25)
  • Sterile Rayon Throat Swabs (25)
  • Positive Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group A Streptococcus
  • Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group C Streptococcus
  • Package Insert (1)
  • Quick Reference Instructions (1)
  • QC Card (located on kit box)
  • Printer Paper (1)
This item is not returnable. View return policy
Description
Description
The Sofia Strep A+ FIA employs immunofluorescence technology that is used with the Sofia Analyzer to detect Group A Streptococcal antigen.
  • For use with Sofia Analyzer
  • CLIA-Waived
  • Highly sensitive and specific with enhanced assay accuracy
  • Analyzes and provides objective result versus visually read tests
  • Involves antigenic components extraction of Group A Streptococcus (GAS) bacteria
  • Results in 5 minutes
Applications:
Healthcare industry for diagnostic purposes
Specifications
Specifications
Room Temperature
Throat Swab
Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
5 min.
Streptococcal Antigen (Group A)
Streptococcus A Starter Kit (CLIA-Waived)
9 Kits (purchase option)