Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel Sofia™ Strep A FIA CLIA-Waived
For rapid qualitative detection of Group A Streptococcal antigens from throat swabs of symptomatic patients
Supplier: Quidel 20280
Includes: 25 Test-Kit
- Individually Packaged Cassettes (25): Polyclonal rabbit anti-Group A Streptococcus antibodies
- Reagent Tubes (25)
- Reagent Solution Bottles (25): 4M Sodium Nitrite and 0.2M Acetic Acid inside glass ampoule
- Fixed Volume Pipettes (25)
- Sterile Rayon Throat Swabs (25)
- Positive Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group A Streptococcus
- Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group C Streptococcus
- Package Insert (1)
- Quick Reference Instructions (1)
- QC Card (located on kit box)
- Printer Paper (1)
Description
The Sofia Strep A+ FIA employs immunofluorescence technology that is used with the Sofia Analyzer to detect Group A Streptococcal antigen.
- For use with Sofia Analyzer
- CLIA-Waived
- Highly sensitive and specific with enhanced assay accuracy
- Analyzes and provides objective result versus visually read tests
- Involves antigenic components extraction of Group A Streptococcus (GAS) bacteria
- Results in 5 minutes
Applications:
Healthcare industry for diagnostic purposes
Order Info
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.
Specifications
Room Temperature | |
Throat Swab | |
Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab | |
5 min. |
Streptococcal Antigen (Group A) | |
Streptococcus A Starter Kit (CLIA-Waived) | |
3 Kits (loaner option) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction