Learn More
Quidel™ QuickVue™ At-Home OTC COVID-19 Self-Test
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The Quickvue™ At-Home OTC COVID-19 test can accurately detect the Omicron variant (among others) of the Sars-CoV-2, the potentially deadly virus that can lead to COVID-19.
$24.49
Specifications
Clia Complexity | Waived (FDA, EUA) |
---|---|
Detectable Analytes | SARS-CoV-2 |
Format | Kit |
Sample Type | Nasal Swab |
Type | At-home COVID Test Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-044-171
![]() |
Quidel™
20402 |
2 Tests/Kit |
Pack of 2 for $24.49
Case of 45 PK for $1,002.00
|
|
|||||
23-044-172
![]() |
Quidel™
20398 |
25 Tests/Kit |
Pack of 25 for $263.00
Case of 10 PK for $2,626.00
|
N/A
![]() |
|||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
QuickVue™ At-Home OTC COVID-19 Test lets you get rapid results, in the privacy of your own home. Available over-the-counter, everything you need is in the package and taking the test is simple.
This home test is authorized for nonprescription home use with self-collected (unobserved) direct anterior nasal (NS) swab specimens from individuals aged 14 years and older or with adult-collected anterior NS samples from individuals aged 2 years or older.
The test to be used in individuals within 6-days of symptom onset or individuals without symptoms or other epidemiological reasons to suspect COVID-19 infection when tested twice over three days with at least 24 hours (and no more than 48 hours) between tests.
The test uses a gentle self-collected anterior nasal (nares) swab sample to determine a positive or negative COVID-19 result. The swab is swirled in a tube of reagent solution, then removed, before a test strip is inserted. After ten minutes, you can take the strip out of the tube and see your results.
Specifications
Waived (FDA, EUA) | |
Kit | |
At-home COVID Test Kit | |
10 mins. |
SARS-CoV-2 | |
Nasal Swab | |
Individually wrapped sterile foam swabs, test strips, single-use strips, pre-filled tubes, tube holder, instruction sheet |
For Use Under an Emergency Use Authorization (EUA) Only. For use with direct anterior nasal swab specimens. For In Vitro Diagnostic Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.