Learn More
Perkin Elmer US LLC Quest ATR Top Plate - ZnSe
Supplier: Perkin Elmer US LLC L1272505
The Quest ATR Accessory is a performance single-reflection ATR accessory designed for sample analysis in the mid- and far-infrared. This accessory has reflective gold-coated optics and Specac's Synopti-Focal Array technology with precision-molded aspheric mirrors. The Quest ATR Accessory is compatible with Spectrum 3/Frontier spectrometers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.