Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Quartzite Specimen (Metamorphic Rock)

For use in comparing with sandstone and metamorphic classification exercises. EISCO Quartzite Specimen (Metamorphic Rock) is formed by applying significant pressure to sandstone. Because of its durable nature, quartzite is often used in paleoclimate studi
$2.80 - $30.75
Specifications
Packaging | Clear labeled and barcoded bag |
---|---|
For Use With (Application) | Comparing with sandstone and metamorphic classification exercises |
Product Type | Quartzite Study Specimen (Metamorphic Rock) |
Width (English) | Approx. 1 in. |
Width (Metric) | Approx. 3 cm |
Description
- Further compaction of sandstone's sturdy silicate framework makes quartzite one of most durable rocks on planet
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- Students can differentiate between quartzite and sandstone by their relative density
- Comes in clear labeled and barcoded bag for storage
Specifications
Clear labeled and barcoded bag | |
Quartzite Study Specimen (Metamorphic Rock) | |
Approx. 3 cm |
Comparing with sandstone and metamorphic classification exercises | |
Approx. 1 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title