Learn More
Chemglass Life Sciences Quartz Wool, Fine, Fiber Size 4-8 Microns
Supplier: Chemglass Life Sciences CGQ069001
Quartz Wool, Fine, Fiber Size 4-8 Microns. Useful as high temperature packing in distillation columns or as a bed support for high purity liquid chromatography. Fine porosity. SOLD IN 1/2 LB. PACKAGES.